Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QPT
Entrez Gene ID
814663
Gene Name
quinolinate phoshoribosyltransferase
Gene Aliases
AT2G01350, F10A8.23, F10A8_23, quinolinate phoshoribosyltransferase
Chromosome Location
Chr.2: 165141 - 167259 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084396.1 NP_001077865.1 2 - 3 443 121
NM_126196.3 NP_565259.1 3 - 4 367 121
NM_201664.2 NP_973393.1 2 - 3 433 121
GenBank mRNA AK117875.1 - 3 - 4 337 121
AY057685.1 - 3 - 4 339 121
AY116955.1 - 3 - 4 317 121

View Full Product Details