Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QPT
Entrez Gene ID
814663
Gene Name
quinolinate phoshoribosyltransferase
Gene Aliases
AT2G01350, F10A8.23, F10A8_23, quinolinate phoshoribosyltransferase
Chromosome Location
Chr.2: 165141 - 167259 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084395.1 NP_001077864.1 4 - 5 487 128
NM_001084396.1 NP_001077865.1 3 - 4 587 128
NM_126196.3 NP_565259.1 4 - 5 511 128
NM_201664.2 NP_973393.1 3 - 4 577 128
GenBank mRNA AK117875.1 - 4 - 5 481 128
AY057685.1 - 4 - 5 483 128
AY116955.1 - 4 - 5 461 128

View Full Product Details