Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl25
Entrez Gene ID
821207
Gene Name
serine carboxypeptidase-like 25
Gene Aliases
AT3G02110, F1C9.10, F1C9_10, serine carboxypeptidase-like 25
Chromosome Location
Chr.3: 370668 - 373805 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111077.4 NP_186860.1 6 - 7 1336 63
GenBank mRNA AY037210.1 - 6 - 7 1223 63
BT002657.1 - 6 - 7 1168 63

View Full Product Details