Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGP1
Entrez Gene ID
821313
Gene Name
UDP-GLUCOSE PYROPHOSPHORYLASE 1
Gene Aliases
AT3G03250, AtUGP1, T17B22.6, T17B22_6, UDP-GLUCOSE PYROPHOSPHORYLASE 1, UDP-glucose pyrophosphorylase, UGP, UGPASE
Chromosome Location
Chr.3: 749309 - 754161 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337461.1 NP_001327658.1 10 - 11 832 127
NM_001337462.1 NP_001327659.1 10 - 11 831 127
NM_111195.4 NP_186975.1 10 - 11 832 127
GenBank mRNA AY035071.1 - 10 - 11 771 127
AY059148.1 - 10 - 11 685 127

View Full Product Details