Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDG8
Entrez Gene ID
821305
Gene Name
homeodomain GLABROUS 8
Gene Aliases
AT3G03260, T17B22.5, T17B22_5, homeodomain GLABROUS 8
Chromosome Location
Chr.3: 755159 - 759282 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337463.1 NP_001326716.1 4 - 5 912 90
NM_111196.2 NP_186976.2 4 - 5 912 90

View Full Product Details