Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPC4
Entrez Gene ID
821243
Gene Name
non-specific phospholipase C4
Gene Aliases
AT3G03530, T21P5.5, T21P5_5, non-specific phospholipase C4
Chromosome Location
Chr.3: 842499 - 845096 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111224.3 NP_566206.1 2 - 3 766 87
GenBank mRNA AB084293.1 - 2 - 3 640 87
AY037257.1 - 2 - 3 679 87
AY143947.1 - 2 - 3 640 87

View Full Product Details