Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAH
Entrez Gene ID
827752
Gene Name
allantoate amidohydrolase
Gene Aliases
AT4G20070, ATAAH, F18F4.170, F18F4_170, allantoate amidohydrolase
Chromosome Location
Chr.4: 10861382 - 10864765 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341398.1 NP_001329687.1 6 - 7 895 124
NM_118126.4 NP_193740.1 6 - 7 899 124
GenBank mRNA BT025334.1 - 6 - 7 733 124

View Full Product Details