Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP71A27
Entrez Gene ID
827771
Gene Name
cytochrome P450, family 71, subfamily A, polypeptide 27
Gene Aliases
''cytochrome P450, AT4G20240, F1C12.160, F1C12.160A, F1C12_160, cytochrome P450, family 71, polypeptide 27, polypeptide 27'', subfamily A
Chromosome Location
Chr.4: 10931335 - 10934237 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341406.1 NP_001329407.1 1 - 2 415 131
NM_118143.4 NP_193757.3 1 - 2 403 131

View Full Product Details