Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATERDJ2B
Entrez Gene ID
827866
Gene Name
DnaJ / Sec63 Brl domains-containing protein
Gene Aliases
AT4G21180, F7J7.120, F7J7_120
Chromosome Location
Chr.4: 11288892 - 11292389 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341475.1 NP_001328817.1 7 - 8 1025 64
NM_118237.4 NP_567621.1 7 - 8 1184 64

View Full Product Details