Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DYT1
- Entrez Gene ID
-
827883
- Gene Name
- basic helix-loop-helix (bHLH) DNA-binding superfamily protein
- Gene Aliases
-
AT4G21330, DYSFUNCTIONAL TAPETUM 1, T6K22.60, T6K22_60
- Chromosome Location
-
Chr.4: 11349922 - 11350694 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118253.1 |
NP_193864.1 |
1 - 2 |
185 |
135 |
View Full Product Details