Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPHK1
Entrez Gene ID
828239
Gene Name
sphingosine kinase 1
Gene Aliases
AT4G21540, F18E5.160, F18E5_160, sphingosine kinase 1
Chromosome Location
Chr.4: 11459228 - 11462651 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341496.1 NP_001320022.1 3 - 4 943 128
NM_118274.8 NP_193885.6 3 - 4 943 128
GenBank mRNA AY128394.1 - 3 - 4 611 128
BT006621.1 - 3 - 4 562 128

View Full Product Details