Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LBD32
Entrez Gene ID
828366
Gene Name
LOB domain-containing protein 32
Gene Aliases
AT4G22700, LOB domain-containing protein 32, T12H17.90, T12H17_90
Chromosome Location
Chr.4: 11933297 - 11933886 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118396.4 NP_194001.1 1 - 1 553 110
GenBank mRNA AB473859.1 - 1 - 1 553 110
DQ056654.1 - 1 - 1 549 110

View Full Product Details