Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ESP
Entrez Gene ID
828396
Gene Name
separase
Gene Aliases
AESP, AT4G22970, EXTRA SPINDLE POLES, F7H19.150, F7H19_150, RADIALLY SWOLLEN 4, RSW4, SEPARASE, homolog of separase
Chromosome Location
Chr.4: 12033437 - 12043728 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203875.2 NP_001190804.1 6 - 7 1771 82
NM_001341570.1 NP_001328112.1 6 - 7 1615 82
NM_118426.4 NP_194028.2 6 - 7 1713 82
GenBank mRNA AY823256.1 - 6 - 7 1615 82

View Full Product Details