The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036624.1 | NP_001031701.1 | 2 - 3 | 518 | 88 |
NM_001203879.2 | NP_001190808.1 | 2 - 3 | 914 | 88 | |
NM_118439.4 | NP_194041.1 | 2 - 3 | 784 | 88 | |
GenBank mRNA | AF419576.1 | - | 2 - 3 | 500 | 88 |
AY143970.1 | - | 1 - 2 | 329 | 88 |