Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSH1
Entrez Gene ID
828409
Gene Name
glutamate-cysteine ligase
Gene Aliases
AT4G23100, ATECS1, AtGSH1, CAD2, CADMIUM SENSITIVE 2, F7H19.290, F7H19_290, GAMMA-GLUTAMYL-CYSTEINE SYNTHETASE, GAMMA-GLUTAMYLCYSTEINE SYNTHETASE, GLUTAMATE-CYSTEINE LIGASE, GSHA, PAD2, PHYTOALEXIN DEFICIENT 2, RML1, ROOT MERISTEMLESS 1, glutamate-cysteine ligase
Chromosome Location
Chr.4: 12102135 - 12107695 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036624.1 NP_001031701.1 6 - 7 944 97
NM_001203879.2 NP_001190808.1 6 - 7 1340 97
NM_118439.4 NP_194041.1 6 - 7 1210 97
GenBank mRNA AF419576.1 - 6 - 7 926 97
AY143970.1 - 5 - 6 755 97

View Full Product Details