Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GH9B15
- Entrez Gene ID
-
828456
- Gene Name
- glycosyl hydrolase 9B15
- Gene Aliases
-
AT4G23560, AtGH9B15, F9D16.30, F9D16_30, glycosyl hydrolase 9B15
- Chromosome Location
-
Chr.4: 12293117 - 12295859 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118487.4 |
NP_194087.1 |
4 - 5 |
752 |
63 |
View Full Product Details