Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CORI3
Entrez Gene ID
828460
Gene Name
Tyrosine transaminase family protein
Gene Aliases
AT4G23600, CORONATINE INDUCED 1, F9D16.70, F9D16_70, JASMONIC ACID RESPONSIVE 2, JR2
Chromosome Location
Chr.4: 12310379 - 12313212 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036628.2 NP_001031705.2 2 - 3 630 78
NM_118491.3 NP_194091.1 2 - 3 870 78
NM_179099.3 NP_849430.1 1 - 2 774 78
GenBank mRNA AF268090.1 - 2 - 3 592 78
AK229608.1 - 1 - 2 404 78
AY099811.1 - 2 - 3 630 78
BT000307.1 - 2 - 3 592 78

View Full Product Details