Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CORI3
Entrez Gene ID
828460
Gene Name
Tyrosine transaminase family protein
Gene Aliases
AT4G23600, CORONATINE INDUCED 1, F9D16.70, F9D16_70, JASMONIC ACID RESPONSIVE 2, JR2
Chromosome Location
Chr.4: 12310379 - 12313212 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036628.2 NP_001031705.2 3 - 4 845 73
NM_118491.3 NP_194091.1 3 - 4 1085 73
NM_179099.3 NP_849430.1 2 - 3 989 73
GenBank mRNA AF268090.1 - 3 - 4 807 73
AK229608.1 - 2 - 3 619 73
AY099811.1 - 3 - 4 845 73
BT000307.1 - 3 - 4 807 73

View Full Product Details