Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPS1
Entrez Gene ID
814888
Gene Name
ureide permease 1
Gene Aliases
AT2G03590, ATUPS1, F19B11.4, F19B11_4, ureide permease 1
Chromosome Location
Chr.2: 1094702 - 1096976 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335192.1 NP_001324930.1 2 - 3 440 69
NM_126409.4 NP_565303.1 1 - 2 655 69
GenBank mRNA AY085962.1 - 1 - 2 427 69
BT029190.1 - 1 - 2 228 69

View Full Product Details