Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPS1
Entrez Gene ID
814888
Gene Name
ureide permease 1
Gene Aliases
AT2G03590, ATUPS1, F19B11.4, F19B11_4, ureide permease 1
Chromosome Location
Chr.2: 1094702 - 1096976 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335192.1 NP_001324930.1 4 - 5 729 95
NM_126409.4 NP_565303.1 3 - 4 944 95
GenBank mRNA AY085962.1 - 3 - 4 716 95
BT029190.1 - 3 - 4 517 95

View Full Product Details