Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAB1
Entrez Gene ID
839871
Gene Name
chlorophyll A/B binding protein 1
Gene Aliases
AB140, AT1G29930, CAB140, CELLULOSE SYNTHASE LIKE C8, CHLOROPHYLL A/B PROTEIN 140, CLSC8, F1N18.3, F1N18_3, LHCB1.3, LIGHT-HARVESTING CHLOROPHYLL A/B-PROTEIN 1.3, chlorophyll A/B binding protein 1
Chromosome Location
Chr.1: 10477885 - 10479114 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102733.3 NP_174286.1 1 - 1 1077 98
GenBank mRNA AF428359.1 - 1 - 1 957 98
AY058180.1 - 1 - 1 957 98

View Full Product Details