Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGR2
Entrez Gene ID
840038
Gene Name
shoot gravitropism 2 (SGR2)
Gene Aliases
AT1G31480, SHOOT GRAVITROPISM 2, T8E3.1, T8E3_1
Chromosome Location
Chr.1: 11265816 - 11271701 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102887.5 NP_174433.2 19 - 20 2622 132
GenBank mRNA AB073133.1 - 19 - 20 2472 132

View Full Product Details