Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AT-IE
Entrez Gene ID
840076
Gene Name
histidine biosynthesis bifunctional protein (HISIE)
Gene Aliases
AT1G31860, F5M6.13, F5M6_13, HISN2, HISTIDINE BIOSYNTHESIS 2, PHOSPHORIBOSYL-ATP PYROPHOSPHOHYDROLASE
Chromosome Location
Chr.1: 11434060 - 11436607 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332984.1 NP_001322577.1 1 - 2 225 78
NM_102923.4 NP_174469.1 1 - 2 225 78
GenBank mRNA AB006082.1 - 1 - 2 193 78
AY046000.1 - 1 - 2 219 78
AY079376.1 - 1 - 2 148 78
AY086514.1 - 1 - 2 225 78

View Full Product Details