The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_120005.4 | NP_195556.1 | 1 - 2 | 524 | 78 |
GenBank mRNA | AY056375.1 | - | 1 - 2 | 345 | 78 |
AY084995.1 | - | 1 - 2 | 441 | 78 | |
AY133649.1 | - | 1 - 2 | 265 | 78 | |
BT000462.1 | - | 1 - 2 | 345 | 78 | |
BT000975.1 | - | 1 - 2 | 265 | 78 |