Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPN10
Entrez Gene ID
830019
Gene Name
regulatory particle non-ATPase 10
Gene Aliases
AT4G38630, ATMCB1, MBP1, MCB1, MULTIUBIQUITIN CHAIN BINDING PROTEIN, MULTIUBIQUITIN CHAIN BINDING PROTEIN 1, MULTIUBIQUITIN-CHAIN-BINDING PROTEIN 1, T9A14.7, regulatory particle non-ATPase 10
Chromosome Location
Chr.4: 18056988 - 18059598 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120024.3 NP_195575.1 9 - 10 1245 107
GenBank mRNA AF360319.1 - 9 - 10 1187 107
AY113889.1 - 8 - 9 1106 107
U33269.1 - 9 - 10 1126 107

View Full Product Details