Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP27
Entrez Gene ID
830092
Gene Name
ubiquitin-specific protease 27
Gene Aliases
AT4G39370, F23K16.5, F23K16_5, ubiquitin-specific protease 27
Chromosome Location
Chr.4: 18306135 - 18308969 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204036.1 NP_001190965.1 2 - 3 655 131
NM_001342538.1 NP_001320167.1 2 - 3 552 131
NM_120097.3 NP_568058.1 2 - 3 586 131
GenBank mRNA AF302675.1 - 2 - 3 416 131
AF370208.1 - 2 - 3 551 131
AY133770.1 - 2 - 3 398 131

View Full Product Details