Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP10
Entrez Gene ID
816141
Gene Name
purple acid phosphatase 10
Gene Aliases
AT2G16430, ATPAP10, F16F14.7, F16F14_7, purple acid phosphatase 10
Chromosome Location
Chr.2: 7120372 - 7123079 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127196.4 NP_179235.1 4 - 5 1032 69
NM_179629.2 NP_849960.1 3 - 4 867 69
GenBank mRNA AF492662.1 - 4 - 5 725 69
AY093236.1 - 4 - 5 773 69
BT008761.1 - 4 - 5 725 69

View Full Product Details