Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP10
Entrez Gene ID
816141
Gene Name
purple acid phosphatase 10
Gene Aliases
AT2G16430, ATPAP10, F16F14.7, F16F14_7, purple acid phosphatase 10
Chromosome Location
Chr.2: 7120372 - 7123079 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127196.4 NP_179235.1 6 - 7 1394 128
NM_179629.2 NP_849960.1 5 - 6 1229 128
GenBank mRNA AF492662.1 - 6 - 7 1087 128
AY093236.1 - 6 - 7 1135 128
BT008761.1 - 6 - 7 1087 128

View Full Product Details