Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCB2;1
Entrez Gene ID
816269
Gene Name
Cyclin B2;1
Gene Aliases
AT2G17620, Cyclin B2;1, T19E12.4, T19E12_4
Chromosome Location
Chr.2: 7664055 - 7666522 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127316.4 NP_179353.1 2 - 3 307 116

View Full Product Details