Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPL1
Entrez Gene ID
819321
Gene Name
squamosa promoter binding protein-like 1
Gene Aliases
AT2G47070, F14M4.10, SQUAMOSA PROMOTER BINDING PROTEIN-LIKE 1, squamosa promoter binding protein-like 1
Chromosome Location
Chr.2: 19336653 - 19340869 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337245.1 NP_001325109.1 10 - 11 2354 67
NM_180137.3 NP_850468.1 9 - 10 2715 67
GenBank mRNA AJ011629.1 - 9 - 10 2268 67
AK230269.1 - 9 - 10 2674 67

View Full Product Details