Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECT1
Entrez Gene ID
819550
Gene Name
evolutionarily conserved C-terminal region 1
Gene Aliases
AT3G03950, T11I18.6, T11I18_6, evolutionarily conserved C-terminal region 1
Chromosome Location
Chr.3: 1021146 - 1024015 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035552.2 NP_001030629.1 3 - 4 699 132
NM_111266.2 NP_566218.1 3 - 4 431 132
NM_180179.3 NP_850510.1 3 - 4 687 132
GenBank mRNA AF419591.1 - 3 - 4 430 132
AK317103.1 - 3 - 4 586 132
AY097342.1 - 3 - 4 331 132
AY894117.1 - 3 - 4 483 132

View Full Product Details