Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRS1
Entrez Gene ID
819671
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
AT3G05090, LATERAL ROOT STIMULATOR 1, T12H1.5, T12H1_5
Chromosome Location
Chr.3: 1417893 - 1423673 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337562.1 NP_001325515.1 13 - 14 1684 105
NM_111381.3 NP_566246.1 13 - 14 1801 105
NM_180185.3 NP_850516.1 13 - 14 1799 105
GenBank mRNA AK226843.1 - 13 - 14 1759 105
AY056292.1 - 13 - 14 1709 105
AY127022.1 - 13 - 14 1752 105
AY142674.1 - 12 - 13 1400 105
BT000631.1 - 12 - 13 1400 105

View Full Product Details