Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRS1
Entrez Gene ID
819671
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
AT3G05090, LATERAL ROOT STIMULATOR 1, T12H1.5, T12H1_5
Chromosome Location
Chr.3: 1417893 - 1423673 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180185.3 NP_850516.1 18 - 19 2670 142
GenBank mRNA AK226843.1 - 18 - 19 2630 142
AY127022.1 - 18 - 19 2623 142

View Full Product Details