Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYSB
Entrez Gene ID
820428
Gene Name
cystatin B
Gene Aliases
ARABIDOPSIS THALIANA PHYTOCYSTATIN 6, AT3G12490, ATCYS6, ATCYSB, cystatin B
Chromosome Location
Chr.3: 3960401 - 3961920 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337993.1 NP_001319530.1 2 - 3 152 64
NM_180239.4 NP_850570.2 1 - 2 241 64

View Full Product Details