Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMC1
Entrez Gene ID
830101
Gene Name
S-adenosylmethionine carrier 1
Gene Aliases
AT4G39460, F23K16.90, F23K16_90, S-adenosylmethionine carrier 1, SAM TRANSPORTER1, SAMT1
Chromosome Location
Chr.4: 18355783 - 18359000 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204039.1 NP_001190968.1 4 - 5 408 127
NM_001342543.1 NP_001328265.1 4 - 5 472 127
NM_120106.3 NP_568060.1 4 - 5 472 127
GenBank mRNA AJ627908.1 - 3 - 4 255 127
AM260490.1 - 3 - 4 255 127
AY046052.1 - 4 - 5 339 127
AY079349.1 - 3 - 4 255 127

View Full Product Details