Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- OSH1
- Entrez Gene ID
-
831721
- Gene Name
- gamma interferon responsive lysosomal thiol (GILT) reductase family protein
- Gene Aliases
-
AT5G01580, F7A7.100, F7A7_100, OAS HIGH ACCUMULATION 1
- Chromosome Location
-
Chr.5: 222760 - 223852 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120236.1 |
NP_195778.1 |
4 - 5 |
539 |
150 |
View Full Product Details