Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHX27
Entrez Gene ID
830460
Gene Name
cation/H+ exchanger 27
Gene Aliases
ARABIDOPSIS THALIANA CATION/HYDROGEN EXCHANGER 27, AT5G01690, ATCHX27, F7A7.210, F7A7_210, cation/H+ exchanger 27
Chromosome Location
Chr.5: 257306 - 260490 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342606.1 NP_001318455.1 4 - 5 2188 95

View Full Product Details