Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPL4
Entrez Gene ID
831758
Gene Name
ubiquitin-protein ligase 4
Gene Aliases
AT5G02880, F9G14.190, F9G14_190, ubiquitin-protein ligase 4
Chromosome Location
Chr.5: 661936 - 669124 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342678.1 NP_001331961.1 7 - 8 1630 131
NM_120366.4 NP_195908.1 7 - 8 1630 131

View Full Product Details