Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATCSLA09
Entrez Gene ID
831734
Gene Name
Nucleotide-diphospho-sugar transferases superfamily protein
Gene Aliases
AT5G03760, ATCSLA9, CELLULOSE SYNTHASE LIKE A9, CSLA09, CSLA9, F17C15.180, F17C15_180, RAT4, RESISTANT TO AGROBACTERIUM TRANSFORMATION 4
Chromosome Location
Chr.5: 985622 - 990630 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120457.4 NP_195996.1 4 - 5 1334 94
GenBank mRNA AY060563.1 - 4 - 5 1254 94
AY149935.1 - 4 - 5 791 94
KJ138709.1 - 4 - 5 791 94

View Full Product Details