Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TPS11
- Entrez Gene ID
-
816385
- Gene Name
- trehalose phosphatase/synthase 11
- Gene Aliases
-
AT2G18700, ATTPS11, ATTPSB, MSF3.8, MSF3_8, TREHALOSE-6-PHOSPHATE SYNTHASE 11, trehalose phosphatase/synthase 11
- Chromosome Location
-
Chr.2: 8108898 - 8112010 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127426.3 |
NP_179460.1 |
1 - 2 |
2106 |
115 |
View Full Product Details