Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- XTH21
- Entrez Gene ID
-
816395
- Gene Name
- xyloglucan endotransglucosylase/hydrolase 21
- Gene Aliases
-
AT2G18800, ATXTH21, MSF3.18, MSF3_18, XYLOGLUCAN ENDOTRANSGLUCOSYLASE/HYDROLASE 21, xyloglucan endotransglucosylase/hydrolase 21
- Chromosome Location
-
Chr.2: 8144881 - 8146477 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127436.2 |
NP_179470.1 |
1 - 2 |
340 |
88 |
View Full Product Details