Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRF1
Entrez Gene ID
816618
Gene Name
STRUBBELIG-receptor family 1
Gene Aliases
AT2G20850, F5H14.18, F5H14_18, STRUBBELIG-receptor family 1
Chromosome Location
Chr.2: 8975313 - 8979437 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335713.1 NP_001324598.1 10 - 11 1401 74
NM_127654.4 NP_565489.2 10 - 11 1401 74

View Full Product Details