Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRT2
Entrez Gene ID
830782
Gene Name
sirtuin 2
Gene Aliases
AT5G09230, AtSRT2, SIRTUIN 2, T2K12.8, sirtuin 2
Chromosome Location
Chr.5: 2871365 - 2873789 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085081.2 NP_001078550.1 10 - 11 1226 77
NM_120959.4 NP_568207.1 10 - 11 1165 77
NM_180464.4 NP_850795.1 10 - 11 1138 77
NM_180465.3 NP_850796.2 9 - 10 1182 77
NM_180466.4 NP_850797.1 9 - 10 1326 77
NM_203024.3 NP_974753.1 9 - 10 1090 77
NM_203025.3 NP_974754.1 9 - 10 1105 77
GenBank mRNA AK316908.1 - 9 - 10 1082 77
AY045873.1 - 10 - 11 1136 77
AY122995.1 - 9 - 10 1066 77

View Full Product Details