Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEP3
Entrez Gene ID
839040
Gene Name
K-box region and MADS-box transcription factor family protein
Gene Aliases
AGAMOUS-like 9, AGL9, AT1G24260, F3I6.19, F3I6_19, SEPALLATA3, TRANSCRIPTION FACTOR AGL9
Chromosome Location
Chr.1: 8593536 - 8596123 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198152.1 NP_001185081.1 6 - 7 867 79
NM_102272.4 NP_564214.2 7 - 8 902 79
NM_180622.3 NP_850953.1 7 - 8 924 79
GenBank mRNA AY088272.1 - 7 - 8 693 79
BT033157.1 - 7 - 8 666 79

View Full Product Details