Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DEK1
Entrez Gene ID
841981
Gene Name
calpain-type cysteine protease family
Gene Aliases
AT1G55350, ATDEK1, DEFECTIVE KERNEL 1, EMB1275, EMB80, EMBRYO DEFECTIVE 80, F7A10.23, F7A10_23, embryo defective 1275
Chromosome Location
Chr.1: 20653740 - 20665110 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198309.1 NP_001185238.1 9 - 10 2493 84
NM_001333706.1 NP_001319240.1 9 - 10 2508 84
NM_001333707.1 NP_001321273.1 9 - 10 2505 84
NM_104411.4 NP_175932.2 9 - 10 2505 84
NM_180635.1 NP_850966.1 8 - 9 2182 84
NM_180636.1 NP_850967.1 8 - 9 2182 84
GenBank mRNA AY061803.1 - 8 - 9 2180 84

View Full Product Details