Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEV1
Entrez Gene ID
830399
Gene Name
Cellulose synthase family protein
Gene Aliases
AT5G05170, ATCESA3, ATH-B, CELLULOSE SYNTHASE 3, CESA3, CONSTITUTIVE EXPRESSION OF VSP 1, ECTOPIC LIGNIFICATION 1, ELI1, ISOXABEN RESISTANT 1, ISOXABEN RESISTANT 1, IXR1, K2A11.4, K2A11_4, MRE1, multiple response expansion 1
Chromosome Location
Chr.5: 1530132 - 1535558 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120599.4 NP_196136.1 11 - 12 2148 110
GenBank mRNA BT002335.1 - 10 - 11 1790 110
KJ138704.1 - 10 - 11 1790 110

View Full Product Details