Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYC1BAT
Entrez Gene ID
830502
Gene Name
Cyclin family protein
Gene Aliases
AT5G06150, CYCB1;2, CYCLIN B1;2, MBL20.2, MBL20_2, cyclin B 1;2
Chromosome Location
Chr.5: 1859279 - 1861703 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120697.3 NP_196233.1 3 - 4 1127 79

View Full Product Details