Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATO
Entrez Gene ID
830503
Gene Name
splicing factor-like protein
Gene Aliases
AT5G06160, ATROPOS, MBL20.3, MBL20_3
Chromosome Location
Chr.5: 1862325 - 1866433 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120698.5 NP_196234.3 10 - 11 1505 76
GenBank mRNA AY039944.1 - 10 - 11 1446 76
AY113862.1 - 10 - 11 1370 76

View Full Product Details