Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIS2
Entrez Gene ID
830553
Gene Name
C2H2 and C2HC zinc fingers superfamily protein
Gene Aliases
AT5G06650, F15M7.18, F15M7_18, GLABROUS INFLORESCENCE STEMS 2
Chromosome Location
Chr.5: 2043419 - 2044200 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120748.2 NP_196283.1 1 - 1 595 57

View Full Product Details