The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_120765.3 | NP_196300.1 | 5 - 6 | 710 | 113 |
GenBank mRNA | FJ708770.1 | - | 5 - 6 | 432 | 113 |